missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethoxyacetophenone, 98%

CAS: 829-20-9 | C10H12O3 | 180.203 g/mol

$130.79 - $280.68

Chemical Identifiers

CAS 829-20-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00008727
InChI Key VQTDPCRSXHFMOL-UHFFFAOYSA-N
Synonym 2',4'-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethanone, 2,4-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 1-2,4-dimethoxyphenyl, acetophenone, 2',4'-dimethoxy, peonol methyl ether, 1-acetyl-2,4-dimethoxybenzene, pubchem10582, acmc-209pqs
PubChem CID 70016
IUPAC Name 1-(2,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1598214
View Documents
Thermo Scientific Chemicals
A1598214
25 g
Each for $130.79
Only null left
 
AAA1598222
View Documents
Thermo Scientific Chemicals
A1598222
100 g
Each for $280.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

829-20-9
180.203
VQTDPCRSXHFMOL-UHFFFAOYSA-N
70016
CC(=O)C1=C(C=C(C=C1)OC)OC
C10H12O3
MFCD00008727
2',4'-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethanone, 2,4-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 1-2,4-dimethoxyphenyl, acetophenone, 2',4'-dimethoxy, peonol methyl ether, 1-acetyl-2,4-dimethoxybenzene, pubchem10582, acmc-209pqs
1-(2,4-dimethoxyphenyl)ethanone

Specifications

829-20-9
287°C to 289°C
C10H12O3
25 g
2',4'-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethanone, 2,4-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 1-2,4-dimethoxyphenyl, acetophenone, 2',4'-dimethoxy, peonol methyl ether, 1-acetyl-2,4-dimethoxybenzene, pubchem10582, acmc-209pqs
CC(=O)C1=C(C=C(C=C1)OC)OC
180.203
180.2
2',4'-Dimethoxyacetophenone
39°C to 41°C
>110°C (230°F)
MFCD00008727
Light sensitive
VQTDPCRSXHFMOL-UHFFFAOYSA-N
1-(2,4-dimethoxyphenyl)ethanone
70016
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-587-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only