missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dimethoxyacetophenone, 98%

Catalog No. AAA1598214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1598214 25 g
AAA1598222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1598214 Supplier Thermo Scientific Chemicals Supplier No. A1598214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 829-20-9 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 829-20-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00008727
InChI Key VQTDPCRSXHFMOL-UHFFFAOYSA-N
Synonym 2',4'-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethanone, 2,4-dimethoxyacetophenone, 1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 1-2,4-dimethoxyphenyl, acetophenone, 2',4'-dimethoxy, peonol methyl ether, 1-acetyl-2,4-dimethoxybenzene, pubchem10582, acmc-209pqs
PubChem CID 70016
IUPAC Name 1-(2,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)OC

Specifications

Melting Point 39°C to 41°C
Boiling Point 287°C to 289°C
Flash Point >110°C (230°F)
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material 2',4'-Dimethoxyacetophenone
EINECSNumber 212-587-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only