missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4'-Dimethoxyacetoacetanilide 95.0+%, TCI America™
$52.19 - $523.79
Chemical Identifiers
| CAS | 16715-79-0 |
|---|---|
| Molecular Formula | C12H15NO4 |
| Molecular Weight (g/mol) | 237.255 |
| MDL Number | MFCD00043929 |
| InChI Key | IQWUCASGTZCNKK-UHFFFAOYSA-N |
| PubChem CID | 85564 |
| IUPAC Name | N-(2,4-dimethoxyphenyl)-3-oxobutanamide |
| SMILES | CC(=O)CC(=O)NC1=C(C=C(C=C1)OC)OC |
Chemical Identifiers
| 16715-79-0 | |
| 237.255 | |
| IQWUCASGTZCNKK-UHFFFAOYSA-N | |
| N-(2,4-dimethoxyphenyl)-3-oxobutanamide |
| C12H15NO4 | |
| MFCD00043929 | |
| 85564 | |
| CC(=O)CC(=O)NC1=C(C=C(C=C1)OC)OC |
Specifications
| 16715-79-0 | |
| C12H15NO4 | |
| 25 g | |
| CC(=O)CC(=O)NC1=C(C=C(C=C1)OC)OC | |
| 237.255 | |
| 237.26 | |
| Crystalline Powder |
| 96°C | |
| MFCD00043929 | |
| IQWUCASGTZCNKK-UHFFFAOYSA-N | |
| N-(2,4-dimethoxyphenyl)-3-oxobutanamide | |
| 85564 | |
| ≥95.0% (N) | |
| 2′,4′-Dimethoxyacetoacetanilide |
Safety and Handling
EINECSNumber : (3)-0740
TSCA : Yes