accessibility menu, dialog, popup

UserName

CAS RN 16715-79-0

IUPAC Name: N-(2,4-dimethoxyphenyl)-3-oxobutanamide
Synonyms: N-(2,4-dimethoxyphenyl)-3-oxobutanamide
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: IQWUCASGTZCNKK-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(=O)CC(C)=O)C(OC)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results