missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxy-beta-methylcinnamic Acid 98.0+%, TCI America™

Catalog No. D50471G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50471G 1 g
D50475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50471G Supplier TCI America Supplier No. D50471G

Chemical Identifiers

CAS 7706-67-4
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00512029
InChI Key VNLOISSPTMDCIF-SOFGYWHQSA-N
Synonym 3-(2,4-Dimethoxyphenyl)-2-butenoic Acid
PubChem CID 6445007
IUPAC Name (E)-3-(2,4-dimethoxyphenyl)but-2-enoic acid
SMILES CC(=CC(=O)O)C1=C(C=C(C=C1)OC)OC

Specifications

Melting Point 158°C
Color White
Quantity 1 g
Formula Weight 222.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxy-beta-methylcinnamic Acid