accessibility menu, dialog, popup

UserName

H3COCH3OHOOH3C(E)

CAS RN 7706-67-4

IUPAC Name: (2E)-3-(2,4-dimethoxyphenyl)but-2-enoic acid
Synonyms: molnia 地美罗酸 dimecrotic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: VNLOISSPTMDCIF-SOFGYWHQSA-N
SMILES: COC1=CC=C(\C(C)=C\C(O)=O)C(OC)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results