missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxy-4'-hydroxybenzophenone 98.0+%, TCI America™

Catalog No. p-7127452
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27075G 5 g
D270725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27075G Supplier TCI America Supplier No. D27075G

Chemical Identifiers

CAS 41351-30-8
Molecular Formula C15H14O4
Molecular Weight (g/mol) 258.273
MDL Number MFCD00137850
InChI Key QEHRETCJMLQPCR-UHFFFAOYSA-N
Synonym 2,4-dimethoxy-4'-hydroxybenzophenone, 2,4-dimethoxyphenyl 4-hydroxyphenyl methanone, 4-2,4-dimethoxybenzoyl phenol, 4'-hydroxy-2,4-dimethoxybenzophenone, 2,4-dimethoxy-4-hydroxybenzophenone, 2,4-dimethoxyphenyl-4-hydroxyphenyl methanone, methanone, 2,4-dimethoxyphenyl 4-hydroxyphenyl, 2,4-dimethoxyphenyl 4-hydroxyphenyl ketone, pubchem7483, acmc-209jk0
PubChem CID 688628
IUPAC Name (2,4-dimethoxyphenyl)-(4-hydroxyphenyl)methanone
SMILES COC1=CC(=C(C=C1)C(=O)C2=CC=C(C=C2)O)OC

Specifications

Melting Point 153°C
Color Red-White
Quantity 5 g
Formula Weight 258.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxy-4′-hydroxybenzophenone