missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxy-1-nitrobenzene 98.0+%, TCI America™
Supplier: TCI America D43825G
Specifications
| 2,4-Dimethoxy-1-nitrobenzene | |
| 76°C | |
| C8H9NO4 | |
| 5 g | |
| XXWIYOBCHKCWNT-UHFFFAOYSA-N | |
| 2,4-dimethoxy-1-nitrobenzene | |
| 78633 | |
| ≥98.0% (GC) |
| 4920-84-7 | |
| Yellow | |
| MFCD00024210 | |
| 1,3-dimethoxy-4-nitrobenzene, 2,4-dimethoxynitrobenzene, 2,4-dimethoxy-1-nitro-benzene, benzene, 2,4-dimethoxy-1-nitro, 2,4-dimethoxylnitrobenzene, pubchem19759, acmc-209kfc, 2,4dimethoxy-nitrobenzene, 2,4-dimethoxy-nitrobenzene, 2.4-dimethoxy-nitrobenzene | |
| COC1=CC(=C(C=C1)[N+](=O)[O-])OC | |
| 183.163 | |
| 183.16 | |
| Crystalline Powder |
Chemical Identifiers
| 4920-84-7 | |
| 183.163 | |
| XXWIYOBCHKCWNT-UHFFFAOYSA-N | |
| 78633 | |
| COC1=CC(=C(C=C1)[N+](=O)[O-])OC |
| C8H9NO4 | |
| MFCD00024210 | |
| 1,3-dimethoxy-4-nitrobenzene, 2,4-dimethoxynitrobenzene, 2,4-dimethoxy-1-nitro-benzene, benzene, 2,4-dimethoxy-1-nitro, 2,4-dimethoxylnitrobenzene, pubchem19759, acmc-209kfc, 2,4dimethoxy-nitrobenzene, 2,4-dimethoxy-nitrobenzene, 2.4-dimethoxy-nitrobenzene | |
| 2,4-dimethoxy-1-nitrobenzene |
Safety and Handling
EINECSNumber : (3)-0801
TSCA : No