missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxyquinoline, 97%

CAS: 86-95-3 | C9H7NO2 | 161.16 g/mol

Supplier:  Thermo Scientific Chemicals B2074714

 View more versions of this product

Catalog No. AAB2074714


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dihydroxyquinoline
>300°C
MFCD00006744
129767
Insoluble in water
C1=CC=C2C(=C1)C(=CC(=O)N2)O
161.16
CHEBI:75926
97%
86-95-3
C9H7NO2
25 g
2,4-quinolinediol, 2,4-dihydroxyquinoline, 4-hydroxyquinolin-2 1h-one, quinoline-2,4-diol, 4-hydroxy-2-quinolone, 4-hydroxycarbostyril, 2 1h-quinolinone, 4-hydroxy, 4-hydroxy-2-quinolinone, hydroxycarbostyril, 2-hydroxyquinolin-4 1h-one
HDHQZCHIXUUSMK-UHFFFAOYSA-N
4-hydroxy-1H-quinolin-2-one
54680871
161.16

Chemical Identifiers

86-95-3
161.16
HDHQZCHIXUUSMK-UHFFFAOYSA-N
54680871
4-hydroxy-1H-quinolin-2-one
C9H7NO2
MFCD00006744
2,4-quinolinediol, 2,4-dihydroxyquinoline, 4-hydroxyquinolin-2 1h-one, quinoline-2,4-diol, 4-hydroxy-2-quinolone, 4-hydroxycarbostyril, 2 1h-quinolinone, 4-hydroxy, 4-hydroxy-2-quinolinone, hydroxycarbostyril, 2-hydroxyquinolin-4 1h-one
CHEBI:75926
C1=CC=C2C(=C1)C(=CC(=O)N2)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 201-711-0

RTECSNumber : FG7300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only