missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxyquinoline, 97%

Catalog No. p-7025323
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2074706 5 g
AAB2074714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2074706 Supplier Thermo Scientific Chemicals Supplier No. B2074706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86-95-3 | C9H7NO2 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86-95-3
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00006744
InChI Key HDHQZCHIXUUSMK-UHFFFAOYSA-N
Synonym 2,4-quinolinediol, 2,4-dihydroxyquinoline, 4-hydroxyquinolin-2 1h-one, quinoline-2,4-diol, 4-hydroxy-2-quinolone, 4-hydroxycarbostyril, 2 1h-quinolinone, 4-hydroxy, 4-hydroxy-2-quinolinone, hydroxycarbostyril, 2-hydroxyquinolin-4 1h-one
PubChem CID 54680871
ChEBI CHEBI:75926
IUPAC Name 4-hydroxy-1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C(=CC(=O)N2)O

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 129767
Solubility Information Insoluble in water
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 2,4-Dihydroxyquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-711-0
RTECSNumber FG7300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only