missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dihydroxypropiophenone 98.0+%, TCI America™

Catalog No. D269425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D269425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D269425G Supplier TCI America Supplier No. D269425G

Chemical Identifiers

CAS 5792-36-9
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002280
InChI Key LLBBBYLDTDJMNU-UHFFFAOYSA-N
Synonym 2',4'-dihydroxypropiophenone, 1-2,4-dihydroxyphenyl propan-1-one, 2,4-dihydroxypropiophenone, 4-propionylresorcinol, 1-propanone, 1-2,4-dihydroxyphenyl, respropiophenone, resorcinol fragment, 9, acmc-209m1c, 2',4'-dihydroxy-propiophenone, 2/',4/'-dihydroxypropiophenone
PubChem CID 79856
IUPAC Name 1-(2,4-dihydroxyphenyl)propan-1-one
SMILES CCC(=O)C1=C(C=C(C=C1)O)O

Specifications

Melting Point 101°C
Boiling Point 178°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,4′-Dihydroxypropiophenone