accessibility menu, dialog, popup

UserName

CH3OOHHO

CAS RN 5792-36-9

IUPAC Name: 1-(2,4-dihydroxyphenyl)propan-1-one
Synonyms: 1-(2,4-dihydroxyphenyl)propan-1-one
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: LLBBBYLDTDJMNU-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(O)C=C1O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)