missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Dihydroxydiphenyl Sulfone 98.0+%, TCI America™

Catalog No. D256725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D256725G 25 g
D2567500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D256725G Supplier TCI America Supplier No. D256725G
Only null left

Chemical Identifiers

CAS 5397-34-2
Molecular Formula C12H10O4S
Molecular Weight (g/mol) 250.27
MDL Number MFCD01631305
InChI Key LROZSPADHSXFJA-UHFFFAOYSA-N
PubChem CID 79381
IUPAC Name 4-(2-hydroxybenzenesulfonyl)phenol
SMILES OC1=CC=C(C=C1)S(=O)(=O)C1=CC=CC=C1O

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 25 g
Formula Weight 250.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4′-Dihydroxydiphenyl Sulfone
EINECSNumber (3)-2169
RTECSNumber SL5365000
TSCA Yes