missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzophenone 98.0+%, TCI America™

Catalog No. p-7126297
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D057325G 25 g
D0573500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D057325G Supplier TCI America Supplier No. D057325G
Only null left

Chemical Identifiers

CAS 131-56-6
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002277
InChI Key ZXDDPOHVAMWLBH-UHFFFAOYSA-N
Synonym 2,4-dihydroxybenzophenone, 2,4-dihydroxyphenyl phenyl methanone, benzophenone-1, benzoresorcinol, resbenzophenone, inhibitor dhbp, uvinul 400, advastab 48, uvistat 12, methanone, 2,4-dihydroxyphenyl phenyl
PubChem CID 8572
ChEBI CHEBI:34240
IUPAC Name (2,4-dihydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)O)O

Specifications

Melting Point 144°C
Color Yellow
Quantity 25 g
Formula Weight 214.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxybenzophenone
EINECSNumber (4)-0129
RTECSNumber DJ0700000
TSCA Yes