missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Dihydroxybenzophenone 98.0+%, TCI America™

Catalog No. D26645G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26645G Supplier TCI America Supplier No. D26645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 606-12-2
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD02181674
InChI Key HUYKZYIAFUBPAQ-UHFFFAOYSA-N
PubChem CID 220295
IUPAC Name (2-hydroxyphenyl)-(4-hydroxyphenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)O)O

Specifications

Melting Point 148°C
Quantity 5 g
Formula Weight 214.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4′-Dihydroxybenzophenone