missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. d0568500g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
D0568500G 500 g
D056825G 25 g
2 options

Catalog No. D0568500G

Supplier: TCI America D0568500G

Only null left

Chemical Identifiers

CAS 89-86-1
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00002451
InChI Key UIAFKZKHHVMJGS-UHFFFAOYSA-N
Synonym beta-resorcylic acid, p-hydroxysalicylic acid, 4-carboxyresorcinol, 4-hydroxysalicylic acid, benzoic acid, 2,4-dihydroxy, beta-resorcinolic acid, 2,4-dhba, resorcylic acid, beta, b-resorcylic acid, 2,4-dihydroxy-benzoic acid
PubChem CID 1491
IUPAC Name 2,4-dihydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)O)C(=O)O

Specifications

Melting Point 218°C
Color White-Yellow
Quantity 500 g
Formula Weight 154.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxybenzoic Acid
EINECSNumber (3)-1616
RTECSNumber VH3708050
TSCA Yes