missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzoic acid, 97%

CAS: 89-86-1 | C7H6O4 | 154.121 g/mol

Supplier:  Thermo Scientific Chemicals A1354536

 View more versions of this product

Catalog No. AAA1354536


Only null left
Explore available promotions

Description

Description

2,4-Dihydroxybenzoic acid is used in Intermediates, Plasticizers, Plastic and Rubber Products, a organic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dihydroxybenzoic acid is used in Intermediates, Plasticizers, Plastic and Rubber Products, a organic building block.

Solubility
Soluble in water(70g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2,4-Dihydroxybenzoic acid
∼226°C (decomposition)
MFCD00002451
1946213
beta-resorcylic acid, p-hydroxysalicylic acid, 4-carboxyresorcinol, 4-hydroxysalicylic acid, benzoic acid, 2,4-dihydroxy, beta-resorcinolic acid, 2,4-dhba, resorcylic acid, beta, b-resorcylic acid, 2,4-dihydroxy-benzoic acid
UIAFKZKHHVMJGS-UHFFFAOYSA-N
2,4-dihydroxybenzoic acid
1491
97%
89-86-1
C7H6O4
500 g
14,8157
Soluble in water(70g/L).
C1=CC(=C(C=C1O)O)C(=O)O
154.121
154.12

Chemical Identifiers

89-86-1
154.121
UIAFKZKHHVMJGS-UHFFFAOYSA-N
1491
C1=CC(=C(C=C1O)O)C(=O)O
C7H6O4
MFCD00002451
beta-resorcylic acid, p-hydroxysalicylic acid, 4-carboxyresorcinol, 4-hydroxysalicylic acid, benzoic acid, 2,4-dihydroxy, beta-resorcinolic acid, 2,4-dhba, resorcylic acid, beta, b-resorcylic acid, 2,4-dihydroxy-benzoic acid
2,4-dihydroxybenzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 201-946-9

RTECSNumber : VH3708050

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only