missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzohydrazide 98.0+%, TCI America™

Catalog No. D47845G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D47845G Supplier TCI America Supplier No. D47845G
Only null left

Chemical Identifiers

CAS 13221-86-8
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00017053
InChI Key GOPUHTXVZGIOHN-UHFFFAOYSA-N
Synonym 2,4-dihydroxybenzhydrazide, 2,4-dihydroxybenzoic acid hydrazide, benzoic acid, 2,4-dihydroxy-, hydrazide, 2,4-dihydroxybenzoic hydrazide, 2,4-bis oxidanyl benzohydrazide, 2,4-dihydroxybenzoic acid, hydrazide, acmc-1cf1y, 4-???benzene-1,3-diol, 2,4-dihydroxybenzoylhydrazine, 2,4-dihydroxy benzoic acid hydrazide
PubChem CID 166785
IUPAC Name 2,4-dihydroxybenzohydrazide
SMILES NNC(=O)C1=CC=C(O)C=C1O

Specifications

Melting Point 236°C
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxybenzohydrazide