missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7126289
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D056425G 25 g
D0564100G 100 g
D0564500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D056425G Supplier TCI America Supplier No. D056425G
Only null left

Chemical Identifiers

CAS 95-01-2
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00011686
InChI Key IUNJCFABHJZSKB-UHFFFAOYSA-N
Synonym beta-resorcylaldehyde, 4-hydroxysalicylaldehyde, 4-formylresorcinol, beta-resorcaldehyde, benzaldehyde, 2,4-dihydroxy, beta-resorcinaldehyde, beta-resorcylic aldehyde, 2,4-dihydroxybenzenecarbonal, salicylaldehyde, 4-hydroxy, 4-hydroxysalicyladehyde
PubChem CID 7213
ChEBI CHEBI:50198
IUPAC Name 2,4-dihydroxybenzaldehyde
SMILES OC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 137°C
Boiling Point 228°C
Quantity 25 g
Formula Weight 138.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxybenzaldehyde
EINECSNumber (3)-1198
RTECSNumber VH3600000
TSCA Yes