accessibility menu, dialog, popup

UserName

OHOHO

CAS RN 95-01-2

IUPAC Name: 2,4-dihydroxybenzaldehyde
Synonyms: 2,4-dihydroxybenzaldehyde β-resorcinaldehyde β-rosorcaldehyde β-resorcylic aldehyde β-resorcylaldehyde β-resorcaldehyde 4-hydroxysalicyladehyde 2,4-dihydroxybenzaldehyd
Molecular Weight (g/mol): 138.12
Molecular Formula: C7H6O3
InChi Key: IUNJCFABHJZSKB-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=O)C(O)=C1
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)