missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. d0564100g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D0564100G 100 g
D056425G 25 g
D0564500G 500 g
3 options

Catalog No. D0564100G

Supplier: TCI America D0564100G

Only null left

Chemical Identifiers

CAS 95-01-2
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00011686
InChI Key IUNJCFABHJZSKB-UHFFFAOYSA-N
Synonym beta-resorcylaldehyde, 4-hydroxysalicylaldehyde, 4-formylresorcinol, beta-resorcaldehyde, benzaldehyde, 2,4-dihydroxy, beta-resorcinaldehyde, beta-resorcylic aldehyde, 2,4-dihydroxybenzenecarbonal, salicylaldehyde, 4-hydroxy, 4-hydroxysalicyladehyde
PubChem CID 7213
ChEBI CHEBI:50198
IUPAC Name 2,4-dihydroxybenzaldehyde
SMILES OC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 137°C
Boiling Point 228°C
Quantity 100 g
Formula Weight 138.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxybenzaldehyde
EINECSNumber (3)-1198
RTECSNumber VH3600000
TSCA Yes