missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzaldehyde, 98%

Catalog No. p-4510588
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173640250 25 g Glass bottle
AC173641000 100 g Glass bottle
AC173645000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC173640250 Supplier Thermo Scientific Chemicals Supplier No. 173640250

CAS: 95-01-2 | C7H6O3 | 138.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-01-2
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00011686
InChI Key IUNJCFABHJZSKB-UHFFFAOYSA-N
Synonym beta-resorcylaldehyde, 4-hydroxysalicylaldehyde, 4-formylresorcinol, beta-resorcaldehyde, benzaldehyde, 2,4-dihydroxy, beta-resorcinaldehyde, beta-resorcylic aldehyde, 2,4-dihydroxybenzenecarbonal, salicylaldehyde, 4-hydroxy, 4-hydroxysalicyladehyde
PubChem CID 7213
ChEBI CHEBI:50198
IUPAC Name 2,4-dihydroxybenzaldehyde
SMILES OC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 133.0°C to 138.0°C
Color White to Beige
Boiling Point 220.0°C to 228.0°C (22.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula (HO)2C6H3CHO
Quantity 25 g
Beilstein 08,241
Merck Index 15,8277
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol,ether,chloroform and,glacial acetic acic
Formula Weight 138.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dihydroxybenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 202-383-1

RUO – Research Use Only