missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dihydroxybenzaldehyde, 98%
Description
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.
Solubility
Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol, ethyl ether, chloroform, glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali.
Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.
Solubility
Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol, ethyl ether, chloroform, glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali.
Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
| Melting Point | 134°C to 138°C |
| Boiling Point | 220°C (22 mmHg) |
| Quantity | 100 g |
| Beilstein | 878548 |
| Sensitivity | Air Sensitive |
| Merck Index | 14,8156 |
| Solubility Information | Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol,ethyl ether,chloroform,glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali. |
| Formula Weight | 138.12 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,4-Dihydroxybenzaldehyde |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 202-383-1 |
| RTECSNumber | VH3600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only