missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dihydroxy-6-methylpyridine 98.0+%, TCI America™
$65.46 - $206.00
Chemical Identifiers
| CAS | 3749-51-7 |
|---|---|
| Molecular Formula | C6H7NO2 |
| Molecular Weight (g/mol) | 125.127 |
| MDL Number | MFCD01860849 |
| InChI Key | WKGSLYHMRQRARV-UHFFFAOYSA-N |
| Synonym | 6-methylpyridine-2,4-diol, 2,4-dihydroxy-6-methylpyridine, 4-hydroxy-6-methylpyridin-2 1h-one, 4-hydroxy-6-methyl-2 1h-pyridinone, 2 1h-pyridinone, 4-hydroxy-6-methyl, 6-methyl-2,4-pyridinediol, 2-hydroxy-6-methyl-1h-pyridin-4-one, 2,4-dihydroxy-6-methyl-pyridine, 4-hydroxy-6-methyl-2-pyridinone, 2 1h-pyridone, 4-hydroxy-6-methyl |
| PubChem CID | 54691419 |
| IUPAC Name | 4-hydroxy-6-methyl-1H-pyridin-2-one |
| SMILES | CC1=CC(=CC(=O)N1)O |
Chemical Identifiers
| 3749-51-7 | |
| 125.127 | |
| WKGSLYHMRQRARV-UHFFFAOYSA-N | |
| 54691419 | |
| CC1=CC(=CC(=O)N1)O |
| C6H7NO2 | |
| MFCD01860849 | |
| 6-methylpyridine-2,4-diol, 2,4-dihydroxy-6-methylpyridine, 4-hydroxy-6-methylpyridin-2 1h-one, 4-hydroxy-6-methyl-2 1h-pyridinone, 2 1h-pyridinone, 4-hydroxy-6-methyl, 6-methyl-2,4-pyridinediol, 2-hydroxy-6-methyl-1h-pyridin-4-one, 2,4-dihydroxy-6-methyl-pyridine, 4-hydroxy-6-methyl-2-pyridinone, 2 1h-pyridone, 4-hydroxy-6-methyl | |
| 4-hydroxy-6-methyl-1H-pyridin-2-one |
Specifications
| 3749-51-7 | |
| White-Yellow | |
| MFCD01860849 | |
| 6-methylpyridine-2,4-diol, 2,4-dihydroxy-6-methylpyridine, 4-hydroxy-6-methylpyridin-2 1h-one, 4-hydroxy-6-methyl-2 1h-pyridinone, 2 1h-pyridinone, 4-hydroxy-6-methyl, 6-methyl-2,4-pyridinediol, 2-hydroxy-6-methyl-1h-pyridin-4-one, 2,4-dihydroxy-6-methyl-pyridine, 4-hydroxy-6-methyl-2-pyridinone, 2 1h-pyridone, 4-hydroxy-6-methyl | |
| CC1=CC(=CC(=O)N1)O | |
| 125.127 | |
| 125.13 | |
| Crystalline Powder |
| 330°C | |
| C6H7NO2 | |
| 1 g | |
| WKGSLYHMRQRARV-UHFFFAOYSA-N | |
| 4-hydroxy-6-methyl-1H-pyridin-2-one | |
| 54691419 | |
| ≥98.0% (GC,T) | |
| 2,4-Dihydroxy-6-methylpyridine |
Safety and Handling
TSCA : No