accessibility menu, dialog, popup

UserName

CH3OHOHN

CAS RN 3749-51-7

IUPAC Name: 2-hydroxy-6-methyl-1,4-dihydropyridin-4-one
Synonyms: 2-hydroxy-6-methyl-1H-pyridin-4-one
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: WKGSLYHMRQRARV-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C=C(O)N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)