missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxy-6-methylpyridine 98.0+%, TCI America™

Catalog No. D43611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43611G 1 g
D43615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43611G Supplier TCI America Supplier No. D43611G

Chemical Identifiers

CAS 3749-51-7
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD01860849
InChI Key WKGSLYHMRQRARV-UHFFFAOYSA-N
Synonym 6-methylpyridine-2,4-diol, 2,4-dihydroxy-6-methylpyridine, 4-hydroxy-6-methylpyridin-2 1h-one, 4-hydroxy-6-methyl-2 1h-pyridinone, 2 1h-pyridinone, 4-hydroxy-6-methyl, 6-methyl-2,4-pyridinediol, 2-hydroxy-6-methyl-1h-pyridin-4-one, 2,4-dihydroxy-6-methyl-pyridine, 4-hydroxy-6-methyl-2-pyridinone, 2 1h-pyridone, 4-hydroxy-6-methyl
PubChem CID 54691419
IUPAC Name 4-hydroxy-6-methyl-1H-pyridin-2-one
SMILES CC1=CC(=CC(=O)N1)O

Specifications

Melting Point 330°C
Color White-Yellow
Quantity 1 g
Formula Weight 125.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxy-6-methylpyridine