missing translation for 'onlineSavingsMsg'
Learn More

2,4-dihydroxy-6-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog No. p-3211520
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC384280050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC384280050 Supplier Thermo Scientific Chemicals Supplier No. 384280050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3749-51-7
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.13
MDL Number MFCD01860849
InChI Key WKGSLYHMRQRARV-UHFFFAOYSA-N
Synonym 6-methylpyridine-2,4-diol, 2,4-dihydroxy-6-methylpyridine, 4-hydroxy-6-methylpyridin-2 1h-one, 4-hydroxy-6-methyl-2 1h-pyridinone, 2 1h-pyridinone, 4-hydroxy-6-methyl, 6-methyl-2,4-pyridinediol, 2-hydroxy-6-methyl-1h-pyridin-4-one, 2,4-dihydroxy-6-methyl-pyridine, 4-hydroxy-6-methyl-2-pyridinone, 2 1h-pyridone, 4-hydroxy-6-methyl
PubChem CID 54691419
IUPAC Name 4-hydroxy-6-methyl-1H-pyridin-2-one
SMILES CC1=CC(=CC(=O)N1)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 125.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 4-Dihydroxy-6-methylpyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only