missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxy-5-methoxypyrimidine 98.0+%, TCI America™

Catalog No. D45991G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45991G 1 g
D45995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45991G Supplier TCI America Supplier No. D45991G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6623-81-0
Molecular Formula C5H6N2O3
Molecular Weight (g/mol) 142.114
MDL Number MFCD00187903
InChI Key KELXHQACBIUYSE-UHFFFAOYSA-N
Synonym 5-Methoxy-2,4-pyrimidinediol
PubChem CID 81100
IUPAC Name 5-methoxy-1H-pyrimidine-2,4-dione
SMILES COC1=CNC(=O)NC1=O

Specifications

Melting Point 344°C
Color White
Quantity 1 g
Formula Weight 142.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxy-5-methoxypyrimidine