accessibility menu, dialog, popup

UserName

CAS RN 6623-81-0

IUPAC Name: 5-methoxy-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-methoxy-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol): 142.11
Molecular Formula: C5H6N2O3
InChi Key: KELXHQACBIUYSE-UHFFFAOYSA-N
SMILES: COC1=CNC(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results