missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4'-Dihydroxy-3'-methylacetophenone, 98%
Description
chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 152°C to 156°C |
| Quantity | 5 g |
| Formula Weight | 166.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 2',4'-Dihydroxy-3'-methylacetophenone |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only