missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dihydroxy-3'-methylacetophenone, 98%

Catalog No. aab2027214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2027214 25 g
AAB2027206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2027214 Supplier Thermo Scientific Chemicals Supplier No. B2027214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10139-84-1 | C9H10O3 | 166.176 g/mol

chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 10139-84-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00010817
InChI Key KMTLZBUHQPQFAV-UHFFFAOYSA-N
Synonym 1-2,4-dihydroxy-3-methylphenyl ethanone, 2',4'-dihydroxy-3'-methylacetophenone, 1-2,4-dihydroxy-3-methylphenyl ethan-1-one, 2,4-dihydroxy-3-methylacetophenone, ethanone, 1-2,4-dihydroxy-3-methylphenyl, acmc-20aocy, 1-acetyl-2,4-dihydroxy-3-methylbenzene, 2 ',4 '-dihydroxy-3 '-methylacetophenone, 1-2,4-dihydroxy-3-methyl-phenyl ethanone, 1-2,4-dihydroxy-3-methyl-phenyl-ethanone
PubChem CID 592139
IUPAC Name 1-(2,4-dihydroxy-3-methylphenyl)ethanone
SMILES CC1=C(C=CC(=C1O)C(=O)C)O

Specifications

Melting Point 152°C to 156°C
Quantity 25 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2',4'-Dihydroxy-3'-methylacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only