missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylhydrazine Hydrochloride 97.0+%, TCI America™

Catalog No. D26985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26985G 5 g
D269825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26985G Supplier TCI America Supplier No. D26985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51523-79-6
Molecular Formula C6H6F2N2
Molecular Weight (g/mol) 144.13
MDL Number MFCD00013383
InChI Key RLZUIPTYDYCNQI-UHFFFAOYSA-N
Synonym 2,4-difluorophenylhydrazine hydrochloride, 2,4-difluorophenyl hydrazine hydrochloride, 2,4-difluorophenylhydrazine hcl, 2,4-difluorohydrazinobenzene hydrochloride, 1-2,4-difluorophenyl hydrazine hydrochloride, 2,4-difluorophenylhydrazinehydrochloride, hydrazine, 2,4-difluorophenyl-, monohydrochloride, 2,4-difluorophenylhydrazine, chloride, pubchem3498, pubchem20774
PubChem CID 2733301
IUPAC Name (2,4-difluorophenyl)hydrazine
SMILES NNC1=CC=C(F)C=C1F

Specifications

Quantity 5 g
Formula Weight 180.58
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Difluorophenylhydrazine Hydrochloride