missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylhydrazine hydrochloride, 97%

Catalog No. p-7081591
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Catalog No. Quantity
AAA1162506 5 g
AAA1162514 25 g
AAA1162522 100 g
3 options

Catalog No. AAA1162506

Supplier: Thermo Scientific Chemicals A1162506

Only null left

CAS: 51523-79-6 | C6H6F2N2 | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51523-79-6
Molecular Formula C6H6F2N2
Molecular Weight (g/mol) 144.13
MDL Number MFCD00013383
InChI Key RLZUIPTYDYCNQI-UHFFFAOYSA-N
Synonym 2,4-difluorophenylhydrazine hydrochloride, 2,4-difluorophenyl hydrazine hydrochloride, 2,4-difluorophenylhydrazine hcl, 2,4-difluorohydrazinobenzene hydrochloride, 1-2,4-difluorophenyl hydrazine hydrochloride, 2,4-difluorophenylhydrazinehydrochloride, hydrazine, 2,4-difluorophenyl-, monohydrochloride, 2,4-difluorophenylhydrazine, chloride, pubchem3498, pubchem20774
PubChem CID 2733301
IUPAC Name (2,4-difluorophenyl)hydrazine;hydrochloride
SMILES NNC1=CC=C(F)C=C1F

Specifications

Melting Point ∼255°C (decomposition)
Quantity 5 g
Beilstein 5411448
Formula Weight 180.59
Percent Purity 98%
Chemical Name or Material 2,4-Difluorophenylhydrazine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only