Learn More
2,4-Difluorophenylhydrazine hydrochloride, 97%
CAS: 51523-79-6 | C6H6F2N2 | 144.13 g/mol
Supplier: Thermo Scientific Chemicals A1162514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,4-Difluorophenylhydrazine hydrochloride | |
| ∼255°C (decomposition) | |
| MFCD00013383 | |
| 5411448 | |
| RLZUIPTYDYCNQI-UHFFFAOYSA-N | |
| (2,4-difluorophenyl)hydrazine;hydrochloride | |
| 2733301 | |
| 98% |
| 51523-79-6 | |
| C6H6F2N2 | |
| 25 g | |
| 2,4-difluorophenylhydrazine hydrochloride, 2,4-difluorophenyl hydrazine hydrochloride, 2,4-difluorophenylhydrazine hcl, 2,4-difluorohydrazinobenzene hydrochloride, 1-2,4-difluorophenyl hydrazine hydrochloride, 2,4-difluorophenylhydrazinehydrochloride, hydrazine, 2,4-difluorophenyl-, monohydrochloride, 2,4-difluorophenylhydrazine, chloride, pubchem3498, pubchem20774 | |
| NNC1=CC=C(F)C=C1F | |
| 144.13 | |
| 180.59 |
Chemical Identifiers
| 51523-79-6 | |
| 144.13 | |
| RLZUIPTYDYCNQI-UHFFFAOYSA-N | |
| 2733301 | |
| NNC1=CC=C(F)C=C1F |
| C6H6F2N2 | |
| MFCD00013383 | |
| 2,4-difluorophenylhydrazine hydrochloride, 2,4-difluorophenyl hydrazine hydrochloride, 2,4-difluorophenylhydrazine hcl, 2,4-difluorohydrazinobenzene hydrochloride, 1-2,4-difluorophenyl hydrazine hydrochloride, 2,4-difluorophenylhydrazinehydrochloride, hydrazine, 2,4-difluorophenyl-, monohydrochloride, 2,4-difluorophenylhydrazine, chloride, pubchem3498, pubchem20774 | |
| (2,4-difluorophenyl)hydrazine;hydrochloride |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only