missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$59.69 - $59.69
Specifications
| CAS | 144025-03-6 |
|---|---|
| Melting Point | 250°C |
| Color | White-Yellow |
| Molecular Formula | C6H5BF2O2 |
| MDL Number | MFCD01318998 |
Specifications
| 144025-03-6 | |
| White-Yellow | |
| MFCD01318998 | |
| (2,4-difluorophenyl)boronic acid | |
| 157.91 |
| 250°C | |
| C6H5BF2O2 | |
| QQLRSCZSKQTFGY-UHFFFAOYSA-N | |
| 2734334 | |
| Crystalline Powder |