missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D33915G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33915G Supplier TCI America Supplier No. D33915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2,4-Difluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 144025-03-6
Melting Point 250°C
Color White-Yellow
Molecular Formula C6H5BF2O2
Quantity 5 g
MDL Number MFCD01318998
Synonym 2,4-difluorobenzeneboronic acid, 2,4-difluorophenyl boronic acid, 2,4-fluorobenzene boronic acid, 2,4-difluorophenyl boranediol, 2,4-difluorophenyl-boronic acid, 2,4-difluorophenylboronicacid, 2 4-difluorophenylboronic acid, boronic acid, 2,4-difluorophenyl, boronic acid, b-2,4-difluorophenyl
InChI Key QQLRSCZSKQTFGY-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(F)C=C1F
IUPAC Name (2,4-difluorophenyl)boronic acid
Molecular Weight (g/mol) 157.91
PubChem CID 2734334
Formula Weight 157.91
Physical Form Crystalline Powder
Show More Show Less