missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylacetone, 98%, Thermo Scientific™

Chemical Identifiers

CAS 274682-91-6
Molecular Formula C9H8F2O
Molecular Weight (g/mol) 170.16
MDL Number MFCD02258866
InChI Key XVHHDTNIOCSTLH-UHFFFAOYSA-N
Synonym 1-2,4-difluorophenyl propan-2-one, 2,4-difluorophenylacetone, 2-propanone, 1-2,4-difluorophenyl, 2,4-difluorophenyl propanone, 1-2,4-difluorophenyl-2-propanone, 1-2,4-difluoro-phenyl-propane-2-one
PubChem CID 2734881
IUPAC Name 1-(2,4-difluorophenyl)propan-2-one
SMILES CC(=O)CC1=C(C=C(C=C1)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC376080010
SDS View Documents
Thermo Scientific Chemicals
376080010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC376080050
SDS View Documents
Thermo Scientific Chemicals
376080050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

274682-91-6
170.16
XVHHDTNIOCSTLH-UHFFFAOYSA-N
2734881
CC(=O)CC1=C(C=C(C=C1)F)F
C9H8F2O
MFCD02258866
1-2,4-difluorophenyl propan-2-one, 2,4-difluorophenylacetone, 2-propanone, 1-2,4-difluorophenyl, 2,4-difluorophenyl propanone, 1-2,4-difluorophenyl-2-propanone, 1-2,4-difluoro-phenyl-propane-2-one
1-(2,4-difluorophenyl)propan-2-one

Specifications

274682-91-6
100.0
97.5% min. (GC)
C9H8F2O
1.4770 to 1.4790
1g
XVHHDTNIOCSTLH-UHFFFAOYSA-N
1-(2,4-difluorophenyl)propan-2-one
2734881
98%
97.5
Authentic
Glass bottle
(F)2C6H3CH2COCH3
MFCD02258866
1-2,4-difluorophenyl propan-2-one, 2,4-difluorophenylacetone, 2-propanone, 1-2,4-difluorophenyl, 2,4-difluorophenyl propanone, 1-2,4-difluorophenyl-2-propanone, 1-2,4-difluoro-phenyl-propane-2-one
CC(=O)CC1=C(C=C(C=C1)F)F
170.16
170.16
2, 4-Difluorophenylacetone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only