accessibility menu, dialog, popup

UserName

CAS RN 274682-91-6

IUPAC Name: 1-(2,4-difluorophenyl)propan-2-one
Synonyms: 1-(2,4-difluorophenyl)propan-2-one
Molecular Weight (g/mol): 170.16
Molecular Formula: C9H8F2O
InChi Key: XVHHDTNIOCSTLH-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results