missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylacetic Acid 98.0+%, TCI America™

Catalog No. D43215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43215G 5 g
D432125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43215G Supplier TCI America Supplier No. D43215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81228-09-3
Molecular Formula C8H5F2O2
Molecular Weight (g/mol) 171.12
MDL Number MFCD00009999
InChI Key QPKZIGHNRLZBCL-UHFFFAOYSA-M
Synonym 2,4-difluorophenylacetic acid, 2-2,4-difluorophenyl acetic acid, benzeneacetic acid, 2,4-difluoro, 2,4-difluorophenyl acetic acid, 2,4-ddifluorophenylacetic acid, rarechem al bo 0247, 2,4-difluoro-phenyl-acetic acid, 2,4-difluorobenzeneacetic acid, 2,4-difluorophenylaceticacid
PubChem CID 123581
IUPAC Name 2-(2,4-difluorophenyl)acetate
SMILES [O-]C(=O)CC1=CC=C(F)C=C1F

Specifications

Melting Point 117°C
Color White
Quantity 5 g
Formula Weight 172.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Difluorophenylacetic Acid