missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenyl Isocyanate 98.0+%, TCI America™

Catalog No. D30865G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D30865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D30865G Supplier TCI America Supplier No. D30865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59025-55-7
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.10
MDL Number MFCD00001997
InChI Key HNENEALJPWJWJY-UHFFFAOYSA-N
Synonym 2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k
PubChem CID 593258
IUPAC Name 2,4-difluoro-1-isocyanatobenzene
SMILES FC1=CC=C(N=C=O)C(F)=C1

Specifications

Color Yellow
Boiling Point 50°C
Quantity 5 g
UN Number 2478
Formula Weight 155.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluorophenyl Isocyanate