missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenyl isocyanate, 98+%

Catalog No. AAA1241506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1241506 5 g
AAA1241514 25 g
AAA1241522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1241506 Supplier Thermo Scientific Chemicals Supplier No. A1241506
Only null left

CAS: 59025-55-7 | C7H3F2NO | 155.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59025-55-7
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.10
MDL Number MFCD00001997
InChI Key HNENEALJPWJWJY-UHFFFAOYSA-N
Synonym 2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k
PubChem CID 593258
IUPAC Name 2,4-difluoro-1-isocyanatobenzene
SMILES FC1=CC=C(N=C=O)C(F)=C1

Specifications

Density 1.31
Boiling Point 64°C to 66°C (10 mmHg)
Flash Point 50°C (122°F)
Refractive Index 1.492
Quantity 5 g
UN Number UN3080
Beilstein 2831577
Sensitivity Moisture sensitive
Formula Weight 155.11
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluorophenyl isocyanate
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H300-H330-H334-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only