missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenol 98.0+%, TCI America™

Catalog No. D18325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18325G 5 g
D183225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18325G Supplier TCI America Supplier No. D18325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 367-27-1
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00009715
InChI Key NVWVWEWVLBKPSM-UHFFFAOYSA-N
Synonym phenol, 2,4-difluoro, 2.4-difluorophenol, 2,4difluorophenol, 2,4-diflurophenol, 2 4-difluorophenol, 2,4 difluorophenol, 2, 4 difluorophenol, 2,4 difluoro phenol, 2,4-di-fluorophenol, 2,4-difluoro phenol
PubChem CID 123051
IUPAC Name 2,4-difluorophenol
SMILES OC1=CC=C(F)C=C1F

Specifications

Color Yellow
Boiling Point 52°C
Quantity 5 g
UN Number 1325
Formula Weight 130.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Difluorophenol