missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenol, 98+%

Catalog No. AAA1404430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1404430 250 g
AAA1404418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1404430 Supplier Thermo Scientific Chemicals Supplier No. A1404430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 367-27-1 | C6H4F2O | 130.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 367-27-1
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00009715
InChI Key NVWVWEWVLBKPSM-UHFFFAOYSA-N
PubChem CID 123051
IUPAC Name 2,4-difluorophenol
SMILES OC1=CC=C(F)C=C1F

Specifications

Melting Point 21°C to 24°C
Density 1.362
Boiling Point 52°C to 54°C (14 mmHg)
Flash Point 56°C (132°F)
Odor Phenol-like
Refractive Index 1.486
Quantity 250 g
UN Number UN2921
Beilstein 1934803
Synonym phenol, 2,4-difluoro, 2.4-difluorophenol, 2,4difluorophenol, 2,4-diflurophenol, 2 4-difluorophenol, 2,4 difluorophenol, 2, 4 difluorophenol, 2,4 difluoro phenol, 2,4-di-fluorophenol, 2,4-difluoro phenol
Formula Weight 130.09
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluorophenol
Show More Show Less
Hazard Category H228-H302+H312+H332-H314
Hazard Statement GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 206-688-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only