missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoronitrobenzene 99.0+%, TCI America™

Catalog No. D163625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D163625G 25 g
D1636500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D163625G Supplier TCI America Supplier No. D163625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 446-35-5
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD00007050
InChI Key RJXOVESYJFXCGI-UHFFFAOYSA-N
Synonym 2,4-difluoronitrobenzene, benzene, 2,4-difluoro-1-nitro, 1-nitro-2,4-difluorobenzene, 1,3-difluoro-4-nitrobenzene, unii-6398y1d7js, 2,4-difluoro-1-nitro-benzene, pubchem2288, 2,4-difluornitrobenzol, 2,4-difluoronitrobenzol, acmc-1akft
PubChem CID 67967
IUPAC Name 2,4-difluoro-1-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(F)C=C1F

Specifications

Boiling Point 207°C
Quantity 25 g
UN Number 2810
Formula Weight 159.09
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluoronitrobenzene
EINECSNumber (3)-0447
RTECSNumber CZ5730000
TSCA Yes