missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Difluorobenzylamine 98.0+%, TCI America™
$32.72 - $67.38
Chemical Identifiers
| CAS | 72235-52-0 |
|---|---|
| Molecular Formula | C7H7F2N |
| Molecular Weight (g/mol) | 143.137 |
| MDL Number | MFCD00010142 |
| InChI Key | QDZZDVQGBKTLHV-UHFFFAOYSA-N |
| Synonym | 2,4-difluorobenzylamine, 2,4-difluorophenyl methanamine, 2,4-difluorobenzyl amine, benzenemethanamine, 2,4-difluoro, 1-2,4-difluorophenyl methanamine, 2,4-difluorophenyl methylamine, n-difluorobenzylamine, pubchem4399, 2,4-difluorbenzylamine, acmc-209oma |
| PubChem CID | 2733244 |
| IUPAC Name | (2,4-difluorophenyl)methanamine |
| SMILES | C1=CC(=C(C=C1F)F)CN |
Chemical Identifiers
| 72235-52-0 | |
| 143.137 | |
| QDZZDVQGBKTLHV-UHFFFAOYSA-N | |
| 2733244 | |
| C1=CC(=C(C=C1F)F)CN |
| C7H7F2N | |
| MFCD00010142 | |
| 2,4-difluorobenzylamine, 2,4-difluorophenyl methanamine, 2,4-difluorobenzyl amine, benzenemethanamine, 2,4-difluoro, 1-2,4-difluorophenyl methanamine, 2,4-difluorophenyl methylamine, n-difluorobenzylamine, pubchem4399, 2,4-difluorbenzylamine, acmc-209oma | |
| (2,4-difluorophenyl)methanamine |
Specifications
| 72235-52-0 | |
| C7H7F2N | |
| 1 g | |
| 2,4-difluorobenzylamine, 2,4-difluorophenyl methanamine, 2,4-difluorobenzyl amine, benzenemethanamine, 2,4-difluoro, 1-2,4-difluorophenyl methanamine, 2,4-difluorophenyl methylamine, n-difluorobenzylamine, pubchem4399, 2,4-difluorbenzylamine, acmc-209oma | |
| C1=CC(=C(C=C1F)F)CN | |
| 143.137 | |
| 143.14 | |
| Liquid |
| 57°C | |
| MFCD00010142 | |
| 2735 | |
| QDZZDVQGBKTLHV-UHFFFAOYSA-N | |
| (2,4-difluorophenyl)methanamine | |
| 2733244 | |
| ≥98.0% (GC) | |
| 2,4-Difluorobenzylamine |
Safety and Handling
TSCA : No