missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzylamine 98.0+%, TCI America™

Catalog No. D19961G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19961G 1 g
D19965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19961G Supplier TCI America Supplier No. D19961G

Chemical Identifiers

CAS 72235-52-0
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.137
MDL Number MFCD00010142
InChI Key QDZZDVQGBKTLHV-UHFFFAOYSA-N
Synonym 2,4-difluorobenzylamine, 2,4-difluorophenyl methanamine, 2,4-difluorobenzyl amine, benzenemethanamine, 2,4-difluoro, 1-2,4-difluorophenyl methanamine, 2,4-difluorophenyl methylamine, n-difluorobenzylamine, pubchem4399, 2,4-difluorbenzylamine, acmc-209oma
PubChem CID 2733244
IUPAC Name (2,4-difluorophenyl)methanamine
SMILES C1=CC(=C(C=C1F)F)CN

Specifications

Boiling Point 57°C
Quantity 1 g
UN Number 2735
Formula Weight 143.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluorobenzylamine