missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzyl Cyanide 98.0+%, TCI America™

Catalog No. D34415G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34415G Supplier TCI America Supplier No. D34415G
Only null left

Chemical Identifiers

CAS 656-35-9
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.132
MDL Number MFCD00009971
InChI Key AGAOESUOSOGZOD-UHFFFAOYSA-N
Synonym 2,4-difluorophenylacetonitrile, 2-2,4-difluorophenyl acetonitrile, 2,4-difluorobenzyl cyanide, 2,4-difluorobenzylcyanide, benzeneacetonitrile, 2,4-difluoro, 2,4-difluorophenyl acetonitrile, 2,4-difluoro-phenyl-acetonitrile, 2-2,4-difluorophenyl ethanenitrile, nc1r bf df
PubChem CID 69565
IUPAC Name 2-(2,4-difluorophenyl)acetonitrile
SMILES C1=CC(=C(C=C1F)F)CC#N

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 g
UN Number 3276
Formula Weight 153.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluorobenzyl Cyanide