missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzenesulfonamide 98.0+%, TCI America™

Catalog No. D51615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51615G 5 g
D516125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51615G Supplier TCI America Supplier No. D51615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13656-60-5
Molecular Formula C6H5F2NO2S
Molecular Weight (g/mol) 193.168
MDL Number MFCD01320743
InChI Key PTHLPYKPCQLAOF-UHFFFAOYSA-N
PubChem CID 574964
IUPAC Name 2,4-difluorobenzenesulfonamide
SMILES C1=CC(=C(C=C1F)F)S(=O)(=O)N

Specifications

Melting Point 156°C
Color White
Quantity 5 g
Synonym 2,4-difluorobenzene-1-sulfonamide, 2,4-difluorobenzenesulphonamide, benzenesulfonamide, 2,4-difluoro, benzenesulfonamide,2,4-difluoro, buttpark 23\07-80, 2,4-difluoro-1-benzenesulfonamide, 2,4-difluorobenzenesulfoneamide, pubchem11764, enamine_001177, acmc-1c82s
Formula Weight 193.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Difluorobenzenesulfonamide