missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoroaniline 98.0+%, TCI America™

Catalog No. D150825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D150825G 25 g
D1508500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D150825G Supplier TCI America Supplier No. D150825G

Chemical Identifiers

CAS 367-25-9
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007648
InChI Key CEPCPXLLFXPZGW-UHFFFAOYSA-N
Synonym 2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine
PubChem CID 9709
IUPAC Name 2,4-difluoroaniline
SMILES NC1=CC=C(F)C=C1F

Specifications

Color Colorless
Boiling Point 170°C
Quantity 25 g
UN Number 2941
Formula Weight 129.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4-Difluoroaniline
EINECSNumber (3)-4289
RTECSNumber BX3680000
TSCA Yes