Learn More
2,4-Difluoroaniline, 99%
CAS: 367-25-9 | C6H5F2N | 129.11 g/mol
Supplier: Thermo Scientific Chemicals A1367314
Description
2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| 2,4-Difluoroaniline | |
| -8°C to -7°C | |
| 168°C to 170°C | |
| Characteristic | |
| 1.507 | |
| 25 g | |
| 2802556 | |
| 2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine | |
| CEPCPXLLFXPZGW-UHFFFAOYSA-N | |
| 2,4-difluoroaniline | |
| 9709 | |
| 99% |
| 367-25-9 | |
| 1.281 | |
| 62°C (143°F) | |
| C6H5F2N | |
| MFCD00007648 | |
| UN2941 | |
| 14,3147 | |
| Not miscible or difficult to mix. | |
| NC1=CC=C(F)C=C1F | |
| 129.11 | |
| 129.11 |
Chemical Identifiers
| 367-25-9 | |
| 129.11 | |
| CEPCPXLLFXPZGW-UHFFFAOYSA-N | |
| 9709 | |
| NC1=CC=C(F)C=C1F |
| C6H5F2N | |
| MFCD00007648 | |
| 2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine | |
| 2,4-difluoroaniline |
Safety and Handling
GHS H Statement
H311-H331-H373-H227-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P311-P312-P330-P361-P363-P370+P378q-P501c
H227-H302-H311+H331
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
EINECSNumber : 206-687-5
RTECSNumber : BX3680000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only